Cole-Parmer® Upright Freezer Drawer Racks for Blood Sample Tubes
Specially designed for storing blood sample tubes
  • Drawers slide out independently for quick access and storage of sample tubes
  • Sturdy corrosion-resistant stainless steel construction for long-term use
Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer.
19,800.00 - ₹58,700.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
1 Drawer, holds (60) 8 to 10 mL tubes 4 1/2 16 1/2
₹19,800.00INR / Each
COMPARE
2 Drawers, holds (120) 8 to 10 mL tubes 9 16 1/2
₹32,000.00INR / Each
COMPARE
2 Drawers, holds (160) 8 to 10 mL tubes 9 22
₹39,100.00INR / Each
COMPARE
2 Drawers, holds (170) 4 to 7 mL tubes 9 16 1/2
₹33,500.00INR / Each
COMPARE
2 Drawers, holds (230) 4 to 7 mL tubes 9 22
₹41,900.00INR / Each
COMPARE
2 Drawers, holds (180) 3 mL tubes 6 1/16 16 1/2
₹38,200.00INR / Each
COMPARE
3 Drawers, holds (375) 3 mL tubes 9 1/8 22
₹58,700.00INR / Each
COMPARE
2 Drawers, holds (324) 1 mL Tubes 6 1/16 22
₹48,500.00INR / Each
Showing 1- 8 of 8