Cole-Parmer® Wide-Mouth Sample Polypropylene Containers
Mix and store in one container
  • Easily attach to blender for in-container mixing
  • Durable for general storage
  • Chemical resistant
  • Autoclavable
Chemical-resistant polypropylene (PP) containers come with white PP linerless lids. Autoclavable.
4,700.00 - ₹12,600.00INR / Pkg of 12
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 43 mm Polypropylene(PP)
₹7,900.00INR / Pkg of 12
COMPARE
60 53 mm Polypropylene(PP)
₹4,700.00INR / Pkg of 12
COMPARE
120 40 mm Polypropylene(PP)
₹8,000.00INR / Pkg of 12
COMPARE
240 89 mm Polypropylene(PP)
₹4,800.00INR / Pkg of 6
COMPARE
480 89 mm Polypropylene(PP)
₹5,900.00INR / Pkg of 6
COMPARE
1000 120 mm Polypropylene(PP)
₹10,500.00INR / Pkg of 6
COMPARE
1200 120 mm Polypropylene(PP)
₹12,600.00INR / Pkg of 6
Showing 1- 7 of 7