Cole-Parmer® Wide Neck Vials
40% larger opening to precisely center a limited-volume insert in the vial neck.
  • Ideal for robotic-arm autosamplers
Choose from clear or amber Type I borosolicate glass. Available with graduated marking spots which can be written on with a #2 pencil.
6,200.00 - ₹6,600.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Outside Diameter (mm) Volume (mL)
Availability Pricing
COMPARE
32 12 2
₹6,200.00INR / Pkg of 100
COMPARE
32 12 2
₹6,600.00INR / Pkg of 100