Corning® Reusable Spinner Flasks
Get better cell production and viability than conventional flasks
  • Side baffles enhance aeration and agitation of flask contents
  • Flasks feature two 45° sidearms
  • Proprietary "stretch" impeller design ensures optimal stirring
  • Autoclavable
This spinner flask offers unsurpassed performance and convenience. All spinner flasks feature unique extended paddle design to ensure uplift flow in all portions of flask providing homogeneous circulation of cells and medium from the bottom of the flask to the top to provide oxygenation and circulation of nutrients. These reusable spinner flasks are equipped with large sidearm openings that easily accept 25- or 50-mL pipettes.
338,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Neck Type Sidearm
Availability Pricing
Corning Reusable Spinner Flask with Angled Sidearms, Glass, 125 mL, 70 mm Center Neck 0183700 Mfr # 4500-125
COMPARE
125 70 mm Center Diameter 32 mm Diameter
Call For Price
Corning Reusable Spinner Flask with Angled Sidearms, Glass, 250 mL, 70 mm Center Neck 0183702 Mfr # 4500-250
COMPARE
250 70 mm Center Diameter 32 mm Diameter
Call For Price
Corning Reusable Spinner Flask with Angled Sidearms, Glass, 500 mL, 100 mm Center Neck 0183704 Mfr # 4500-500
COMPARE
500 100 mm Center Diameter 45 mm Diameter
Call For Price
Corning Reusable Spinner Flask with Angled Sidearms, Glass, 1000 mL, 100 mm Center Neck 0183706 Mfr # 4500-1L
COMPARE
1000 100 mm Center Diameter 45 mm Diameter
Call For Price
Corning Reusable Spinner Flask with Angled Sidearms, Glass, 3000 mL, 100 mm Center Neck 0183708 Mfr # 4500-3L
COMPARE
3000 100 mm Center Diameter 45 mm Diameter
Call For Price
Corning Reusable Spinner Flask with Angled Sidearms, Glass, 3000 mL, 120 mm Center Neck 0183710 Mfr # 4502-3L
COMPARE
3000 120 mm Center Diameter 45 mm Diameter
₹338,000.00INR / Each