Corning Single Use Sterile Spatulas, PS
  • Modified polystyrene construction—nontoxic and chemical resistant
Never waste time resterilizing spatulas again. Each spatula is individually packaged and comes gamma irradiation sterilized for no ETO residues. Spatulas are RNase/DNase free, nonpyrogenic, and antistatic.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Length (in) Sterile Description
Availability Pricing
Corning 3003 Single-use Sterile spatula, tapered blade/spoon 0626500 Mfr # 3003
COMPARE
9 11/16 Yes Single-use Sterile spatula, tapered blade/spoon
Call For Price
Corning 3004 Single-use Sterile spatula, small spoon/spoon 0626502 Mfr # 3004
COMPARE
9 11/16 Yes Single-use Sterile spatula, small spoon/spoon
Call For Price
Corning 3005 Single-use Sterile spatula, round end/spoon 0626504 Mfr # 3005
COMPARE
9 11/16 Yes Single-use Sterile spatula, round end/spoon
Call For Price
Corning 3006 Single-use Sterile spatula, V-scoop/spoon 0626506 Mfr # 3006
COMPARE
9 11/16 Yes Single-use Sterile spatula, V-scoop/spoon
Call For Price
Corning 3007 Single-use Sterile spatula, flat end/spoon 0626508 Mfr # 3007
COMPARE
9 11/16 Yes Single-use Sterile spatula, flat end/spoon