Diba Omnifit® PTFE Tubing
Chemically inert tubing withstands most chemicals, solvents, and corrosive materials
  • Constructed of durable PTFE
Use chemically inert PTFE tubing with virtually all chemicals, solvents, and corrosive materials, even at elevated temperatures. Autoclave tubing or sterilize in-line by steam or chemical methods. Select from 1/16", 1/8", or 3/16" OD tubing with a range of inside diameters.
12,600.00 - ₹305,000.00INR / Pkg of 1
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Length (m) Tubing ID (mm) Tubing OD (mm)
Availability Pricing
Diba Omnifit® Tubing, PTFE, 1/16" (1.6 mm) OD x 0.3 mm ID; 20 m/pk 2194268 Mfr # 008T16-030-20
COMPARE
20 0.3 1.6
₹12,600.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 1/16" (1.6 mm) OD x 0.3 mm ID; 200 m/pk 2194269 Mfr # 008T16-030-200
COMPARE
200 0.3 1.6
₹115,000.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 1/16" (1.6 mm) OD x 0.5 mm ID; 20 m/pk 2194270 Mfr # 008T16-050-20
COMPARE
20 0.5 1.6
₹12,600.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 1/16" (1.6 mm) OD x 0.5 mm ID; 200 m/pk 2194271 Mfr # 008T16-050-200
COMPARE
200 0.5 1.6
₹115,000.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 1/16" (1.6 mm) OD x 0.8 mm ID; 20 m/pk 2194272 Mfr # 008T16-080-20
COMPARE
20 0.8 1.6
₹12,600.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 1/16" (1.6 mm) OD x 0.8 mm ID; 200 m/pk 2194273 Mfr # 008T16-080-200
COMPARE
200 0.8 1.6
₹115,000.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 1/16" (1.6 mm) OD x 1.0 mm ID; 20 m/pk 2194274 Mfr # 008T16-100-20
COMPARE
20 1 1.6
₹12,600.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 1/16" (1.6 mm) OD x 1.0 mm ID; 200 m/pk 2194275 Mfr # 008T16-100-200
COMPARE
200 1 1.6
₹112,000.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 1/8" (3.2 mm) OD x 1.5 mm ID; 10 m/pk 2194276 Mfr # 008T32-150-10
COMPARE
10 1.5 3.2
₹16,200.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 1/8" (3.2 mm) OD x 1.5 mm ID; 100 m/pk 2194277 Mfr # 008T32-150-100
COMPARE
100 1.5 3.2
₹151,000.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 3/16" (4 mm) OD x 3.2 mm ID; 10 m/pk 2194278 Mfr # 008T47-032-10
COMPARE
10 3.2 4.8
₹31,000.00INR / Pkg of 1
Diba Omnifit® Tubing, PTFE, 3/16" (4 mm) OD x 3.2 mm ID; 100 m/pk 2194279 Mfr # 008T47-032-100
COMPARE
100 3.2 4.8
₹305,000.00INR / Pkg of 1
Showing 1- 12 of 12