Digi-Sense AM1610 Series-Precision Industrial PRT / RTD Probes
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
AM1610 Series—Precision industrial PRT/RTD probes: Conform to the standard 385 curve, letting you use your DIN/IEC RTD meters fully. Manufactured using a special aged and tested 5mm long sensing element to achieve the best performance in stability and repeatability. All have an accuracy of 35 mK at 0°C and short-term stability of 10 mK at 0°C. The AM1610 has a temperature range of -60 to 160ºC (-76 to 320ºF)
108,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
Stripped Wire 320 -76
₹108,000.00INR / Each
COMPARE
Spade Lugs 320 -76
₹108,000.00INR / Each
COMPARE
Stripped Wire 320 -76
₹108,000.00INR / Each