Digi-Sense AM1612 Series Full Immersion PRT / RTD Probes
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5606
AM1612 and AM1642 Series—Full-immersion PRT/RTD probes: Designed to perform in extreme environments where both the transition junction and the lead wires are required to withstand temperatures covering the entire operating range of the probe. Applications include calibration or validation of sensors used in laboratory or biology freezers, walk-in refrigerators, autoclaves, ovens, stability test chambers, furnaces, and incubators. These probes feature short term stability of 20 mK at TPW and long term drift of 40 mK at Triple Point of Water (TPW). The AM1612 has a temperature range of -196 to 180ºC (-320.8 to 356ºF)
139,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
COMPARE
Stripped Wire 356
₹139,000.00INR / Each
COMPARE
Spade Lugs 356
₹139,000.00INR / Each
COMPARE
5-Pin DIN 356
₹139,000.00INR / Each