Digi-Sense AM1730 Series-Secondary Reference PRT / RTD Probes
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5615 and 5616
AM1730—Secondary reference PRT/RTD probes: These probes are the workhorse of laboratories. With a calibration uncertainty at 0°C of just ±0.015°C, these probes fully meet ITS-90 criteria for reference thermometers fully. The unique design achieves an excellent balance between hysteresis effect and mechanical/thermal shocks. Both series boast very low short term stability of 7 mK at TPW. The AM1730 has a range of -200 to 420°C.
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.
225,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
Stripped Wire 788 -328
₹225,000.00INR / Each
COMPARE
5-Pin DIN 788 -328
₹225,000.00INR / Each
COMPARE
Stripped Wire 572 -76
₹225,000.00INR / Each
COMPARE
Spade Lugs 572 -76
₹225,000.00INR / Each