Digi-Sense® Blue Spirit Laboratory Standard Accuracy Glass Thermometers, Celsius-scale
Scales are permanently fused on to the glass and won’t wear off
  • Nonmercury blue spirit liquid
  • Statement of accuracy
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Choose from partial immersion or total immersion thermometers.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.
6,200.00 - ₹7,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Immersion Depth (mm) Max Temperature (° C) Min Temperature (° C)
Availability Pricing
COMPARE
50 110 -10
Call For Price
COMPARE
50 150 -10
Call For Price
COMPARE
76 110 -20
Call For Price
COMPARE
76 260 -10
Call For Price
COMPARE
200 110 -10
Call For Price
COMPARE
300 110 -20
Call For Price
COMPARE
300 150 -20
Call For Price
COMPARE
350 225 -10
Call For Price
COMPARE
76 50 -50
₹6,900.00INR / Each
COMPARE
76 50 -100
₹6,800.00INR / Each
COMPARE
300 50 -50
₹6,200.00INR / Each
COMPARE
300 50 -100
₹7,000.00INR / Each
Showing 1- 12 of 12