Digi-Sense RTD Bendable-Seath Probes
RTD Bendable Probes are Designed to Measure Temperature in a Confined Space
  • The first 3” of the probe tip are not to be bent.
  • Time Constant: 3 seconds or greater
  • Custom designs or changes to this product are available upon request.
Maximum error for probe assembly is ±1.1°C + 0.12% of reading below 300°F, ±0.35% of reading above 300°F. Alpha coefficient is 0.003850 omega/omega/°C (DIN IEC 751). Resistance at ice point (0°C) is 100 omega.
Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
15,000.00 - ₹24,900.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
COMPARE
Digi-Sense 3-pin 900
₹15,000.00INR / Each
COMPARE
Digi-Sense 3-pin 932
₹20,400.00INR / Each
COMPARE
Digi-Sense 3-pin 932
₹22,400.00INR / Each
COMPARE
Digi-Sense 3-pin 932
₹24,400.00INR / Each
COMPARE
Spade Lugs 932
₹21,700.00INR / Each
COMPARE
Spade Lugs 932
₹22,000.00INR / Each
COMPARE
ANSI 3-blade 932
₹22,400.00INR / Each
COMPARE
ANSI 3-blade 932
₹23,200.00INR / Each
COMPARE
ANSI 3-blade 932
₹24,900.00INR / Each
Showing 1- 9 of 9