Digi-Sense Temperature Probe Thermocouple Feedthroughs Wire Assembles
Use these two-wire feedthroughs to make connections inside vessels or chambers. Use with probes with miniconnectors. Feedthroughs withstand vacuum conditions or pressures to 100 psi. Stainless steel and epoxy construction. Temperature range: -40 to 250°> F (-40 to 121°> C). Feedthroughs have a 1/2'' NPT(M) fitting. Include 48''L, PTFE insulated, 24-gauge (0.0201'' wire dia) lead wires.
16,000.00 - ₹16,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connector Type Length (in) Use With Probe / Wire Type
Availability Pricing
COMPARE
₹16,600.00INR / Each
COMPARE
₹16,000.00INR / Each