Digi-Sense Thermocouple Alligator Clip Surface Probes for Ovens
Ideal for permanent installation inside ovens or other hot dry environments
  • The exposed junction is inside the clip for fast response
  • The alligator clip is 2” long with a 5/16” max jaw opening
  • Response Time: 2.25 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
10,300.00 - ₹10,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
400 J
₹10,300.00INR / Each
COMPARE
650 K
₹10,900.00INR / Each
COMPARE
400 T
₹10,300.00INR / Each