Digi-Sense Thermocouple Flexible Wire Probes - FEP-Insulated with Epoxy-Coated Junction
Insulated with FEP-Sealed Junction Used for Long-Term Immersion in Semi-Caustic Environments
  • Each Probe is hermetically sealed and tested to 25psi
  • Can be used for applications requiring an isolated junction.
  • Response Time: 0.75 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
  • Custom designs or changes to this product are available upon request
Do not subject type J exposed-junction probes to moisture as they will rust
5,900.00 - ₹6,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Description
Availability Pricing
COMPARE
400 J Type-J, FEP-Insulated Probe, Coated Junct, Mini Conn, Ungrd, 10ft 24-Gauge
₹5,900.00INR / Each
COMPARE
400 K Type-K, FEP-Insulated Probe, Coated Junct, Mini Conn, Ungrd, 10ft 24-Gauge
₹6,100.00INR / Each
COMPARE
400 T Type-T, FEP-Insulated Probe, Coated Junct, Mini Conn, Ungrd, 10ft 24-Gauge
₹6,100.00INR / Each