Digi-Sense Thermocouple General-Purpose Stainless Steel Probes
Gerneral-Purpose Stainless Steel Probes Are Designed For Use with Liquids, Gases and Semisolids.
  • Ideal For Checking Food Temperatures
  • Response Time: 2 seconds or greater.
  • Durable for heavy duty use or industrial applications.
  • Stainless Steel handles provide use in higher temperatures and increased durability.
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
19,400.00 - ₹20,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
1000 J
₹19,400.00INR / Each
COMPARE
1200 K
₹19,500.00INR / Each
COMPARE
600 T
₹20,100.00INR / Each