Digi-Sense Thermocouple High-temperature Probes
High-Temperature Thermocouple Probes Designed for Multiple Bends Ideal for any Environment
  • Bend probes around pipe diameters as small as twice the sheath diameter to easily determine your pipe temperature
  • Ideal for industrial environments where fast accurate readings are required
  • Response Time: .85 seconds or greater
  • Type “K” and “J” probes feature ICONEL 600 sheaths
  • Type “T” probes feature 316 Stainless Steel sheaths
  • Fiberglass insulated cable with stainless steel overbraid
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
6,200.00 - ₹15,100.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Sheath Length (in)
Availability Pricing
Digi-Sense Type T High-Temperature Thermocouple Probes; 12" L 0850096
₹6,200.00INR / Each
COMPARE
750 T 12
Loading
₹6,200.00INR / Each
Digi-Sense Type E High-Termperature Thermocouple Probes; 12" L 0851296
₹6,600.00INR / Each
COMPARE
1800 E 12
Loading
₹6,600.00INR / Each
Digi-Sense Type K High-Temperature Flexible Thermocouple Probe; 12" L 0851696
₹7,500.00INR / Each
COMPARE
2012 K 12
Loading
₹7,500.00INR / Each
Digi-Sense Type K High-Temperature Thermocouple Probes; 25" L 0851697
₹15,100.00INR / Each
COMPARE
2012 K 25
Loading
₹15,100.00INR / Each
Digi-Sense Type J High-Temperature Thermocouple Probes; 12" L 0851796
₹7,000.00INR / Each
COMPARE
500 J 12
Loading
₹7,000.00INR / Each
Digi-Sense Type J High-Temperature Thermocouple Probes; 25" L 0851797
₹12,800.00INR / Each
COMPARE
500 J 25
Loading
₹12,800.00INR / Each