Digi-Sense Thermocouple Pipe-Fitting Probes
Thermocouple Pipe-Fitting Probes Ideal for High Temperature Environments or Pressurized Chambers
  • Each Probe comes with a one-time adjustable stainless steel compression fitting with a 1/8” NPT process.
  • Compression fittings can hold up to 2000psi once installed
  • Response Time: 3 seconds or greater
  • Fiberglass insulated cable ideal for continuous high temperature environments
  • Type “K” and “J” probes feature ICONEL 600 sheaths
  • Type “T” probes feature 316 Stainless Steel sheaths
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
8,400.00 - ₹10,200.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Description
Availability Pricing
COMPARE
500 T Type-T Pipe-ftg Probe SS Handle Mini Conn 12" L .188" Dia Grd 6ft Cable
₹10,200.00INR / Each
COMPARE
500 T Type-T Pipe-ftg Probe SS Handle Mini Conn 4" L .188" Dia Grd 6ft Cable
₹8,400.00INR / Each
COMPARE
2012 K Type-K Pipe-ftg Probe SS Handle Mini Conn 12" L .188" Dia Grd 6ft Cable
₹10,100.00INR / Each
COMPARE
2012 K Type-K Pipe-ftg Probe SS Handle Mini Conn 4" L .188" Dia Grd 6ft Cable
₹8,500.00INR / Each
COMPARE
1600 J Type-J Pipe-ftg Probe SS Handle Mini Conn 12" L .188" Dia Grd 6ft Cable
₹10,100.00INR / Each
COMPARE
1600 J Type-J Pipe-ftg Probe SS Handle Mini Conn 4" L .188" Dia Grd 6ft Cable
₹8,900.00INR / Each