Digi-Sense Thermocouple Wire - 16 Gauge
Make your own extension cables
Wire comes in 16-gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with polyvinyl chloride (PVC), fluorinated ethylene propylene resin (FEP/FEP), fiberglass, high-temperature fiberglass, or ceramic fiber outer/inner insulation. These cables are also available in 100-ft or 1000-ft lengths for any application.
20,600.00 - ₹509,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Material Max Temperature (° F)
Availability Pricing
COMPARE
100 Polyvinyl chloride (PVC) 221
₹20,600.00INR / Each
COMPARE
100 Polyvinyl chloride (PVC) 221
₹36,500.00INR / Each
COMPARE
100 Fiberglass 900
₹50,900.00INR / Each
COMPARE
1000 Fiberglass 900
₹509,000.00INR / Each