Drager Diffusion Tubes
Direct read tubes identify workers exposure to chemicals
  • Read exposure levels easily but he markings on the side of the tube
Direct-reading diffusion tubes are used specifically for personal exposure monitoring. These tubes do not require the use of a gas detector pump because the contaminant molecules pass into the tube by diffusion processes and in this manner are brought into contact with the reagent systems. Exposure levels are easily read by the markings on the side of the tube.
28,300.00 - ₹46,000.00INR / Pkg of 3
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Gas Measured Range Description
Availability Pricing
Drager Diffusion Tube Holder with clip, pack of 3 8651550 Mfr # 6733014
₹28,300.00INR / Pkg of 3
COMPARE
Diffusion Tube Holder with clip, pack of 3
Loading
₹28,300.00INR / Pkg of 3
Drager Diffusion Tube, Ammonia 20/a-D, 20 to 1500 ppm at 1 hour 8651552 Mfr # 8101301
₹46,000.00INR / Box of 10
COMPARE
Ammonia 20/a-D 1 hr: 20 to 1500 ppm, 4 hr: 2.5 to 200 ppm Diffusion Tube, Ammonia 20/a-D, 20 to 1500 ppm at 1 hour
Loading
₹46,000.00INR / Box of 10
Drager Diffusion Tube Nitrogen Dioxide 10/a-D 8651561 Mfr # 8101111
₹46,000.00INR / Box of 10
COMPARE
Nitrogen dioxide 10/a-D 1 hr: 10 to 200 ppm, 4 hr: 1.3 to 25 ppm Diffusion Tube Nitrogen Dioxide 10/a-D
Loading
₹46,000.00INR / Box of 10