EM Quant Water Test Strips

Use for quick spot checks to ensure water quality
  • Convenient, on-site water testing
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.
33,600.00 - ₹36,900.00INR / Pkg of 100

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Measurement Points
Availability Pricing
EMD Millipore 1.10007.0001 Nitrite Test Strips, 2 to 80 ppm, 100/Pk 9957215 Mfr # 1.10007.0001
COMPARE
0 to 80 ppm
₹36,900.00INR / Pkg of 100
EMD Millipore 1.10013.0001 Sulfite Test Strips, 10 to 400 ppm, 100/Pk 9957216 Mfr # 1.10013.0001
COMPARE
0 to 400 ppm
₹33,600.00INR / Pkg of 100
EMD Millipore 1.10019.0001 Sulfate Test Strips, 200 to 1600 ppm, 100/Pk 9957217 Mfr # 1.10019.0001
COMPARE
200 to 1600 ppm
₹35,400.00INR / Pkg of 100
EMD Millipore 1.17953.0001 Zinc Test Strips, 0 to 50 mg/L, 100/Pk 9957218 Mfr # 1.17953.0001
COMPARE
0 to 50 ppm
Call For Price
EMD Millipore 1.17985.0001 Potassium Test Strips, 100 9957219 Mfr # 1.17985.0001
COMPARE
0 to 1500 ppm
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat