Environmental Express® FilterMate™ Accessories
No need for a brand new FilterMate
  • Plunger or sample tube available
  • Works with 50 mL sample tube and FilterMate
The machined HDPE holder adds stability when performing FilterMate filtrations. The 50 mL sample tube is placed into the center hole of the FilterMate holder prior to filtration. The holder secures the digestion tube as the FilterMate is depressed through the liquid sample.
17,500.00 - ₹31,300.00INR / Pkg of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Environmental Express FilterMate™ 50 mL Digestion Tube Holder 3520206
₹17,500.00INR / Pkg of 1
COMPARE
50 mL Digestion Tube Holder
Loading
₹17,500.00INR / Pkg of 1
Environmental Express FilterMate™ Replacement Plungers, 50 mL; 500/Pk 3520207
₹31,300.00INR / Pkg of 500
COMPARE
Replacement Plungers, 50 mL; 500/Pk
Loading
₹31,300.00INR / Pkg of 500