Environmental Express® FilterMate Systems
Convenient filtration without a transfer step
  • Simple to use just press on plunger
  • Filter 15- and 50- mL samples within sample tubes
  • No need for separate filtration devices before analyzing samples
Soil, sludge, wastewater, and challenging samples require filtration prior to analysis. FilterMate fits directly in the digestion vessel and is pushed through the liquid being filtered to the bottom of the tube with a detachable plunger. After filtration, the plunger is easily removed and discarded while the filter assembly remains in the bottom of the tube. Finished sample can be capped for storage or poured directly into an autosampler vial.
14,500.00 - ₹47,500.00INR / Pkg of 100
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Funnel Size (mL) Pore Size (µm)
Availability Pricing
COMPARE
PTFE 50 5
₹22,600.00INR / Pkg of 100
COMPARE
Glass 50 0.7
₹28,600.00INR / Pkg of 100
COMPARE
PVDF with PTFE prefilter 50 0.45
₹33,700.00INR / Pkg of 100
COMPARE
Certified PTFE 50 6
₹28,100.00INR / Pkg of 100
COMPARE
Certified PVDF with PTFE prefilter 50 0.45
₹47,500.00INR / Pkg of 100
COMPARE
Polyethylene (PE) 15
₹14,500.00INR / Pkg of 100