Environmental Express® FlipMate Filtration Assemblies
Assemblies connect directly to your existing digestion tubes
  • Sample is ready in just seconds
  • Convenient, economical method for filtration
  • Produce quality results with clean samples
The FlipMate assemblies easily filter your sample with the use of two threaded digestion tubes, a little tubing, and vacuum pressure. The sample is placed in one tube, the filter assembly threaded onto the top of the tube, then the tube and filter are flipped, the receiving tube attached, vaccum tubing attached, vaccum pressure applied, and the sample is then pulled from the sample tube, through the filter assembly, and into the empty tube.
17,500.00 - ₹45,600.00INR / Pkg of 100
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Funnel Size (mL) Pore Size (µm)
Availability Pricing
COMPARE
PES 50 0.1
₹17,500.00INR / Pkg of 100
COMPARE
PES with PTFE pre-filter 50 0.45
₹26,200.00INR / Pkg of 100
COMPARE
Glass without pre-filter 50 1.5
₹25,100.00INR / Pkg of 100
COMPARE
PES without pre-filter 50 0.45
₹24,000.00INR / Pkg of 100
COMPARE
Certified PES with PTFE pre-filter 50 0.45
₹45,600.00INR / Pkg of 100
COMPARE
Glass with PTFE pre-filter 50 0.7
₹25,100.00INR / Pkg of 100
COMPARE
Acid Washed Glass without pre-filter 50 0.7
₹25,100.00INR / Pkg of 100
COMPARE
Certified, PVDF without pre-filter 50 0.45
₹34,600.00INR / Pkg of 100
COMPARE
PES with PTFE pre-filter 100 0.45
₹19,400.00INR / Pkg of 50
COMPARE
Glass without pre-filter 100 1.5
₹19,600.00INR / Pkg of 50
COMPARE
PES with PTFE pre-filter 100 0.2
₹19,700.00INR / Pkg of 50
COMPARE
Acid-washed glass 100 0.7
₹19,400.00INR / Pkg of 50
COMPARE
Acid-washed glass 100 10
Call For Price
COMPARE
Certified PES with PTFE pre-filter 100 0.45
₹32,700.00INR / Pkg of 50
Showing 1- 14 of 14