Erlab CaptairFlow Clean Air Enclosures
Provides an ultra-clean, dust-free work area
  • HEPA filter provides clean workstation air
  • UV lights help decontaminate worktop
The Clean Air Enclosures are designed to prevent environmental pollution and cross-contamination for applications such as non-pathogenic cell cultures, in-vitro cultures, microbiology, homeopathic preparations, electronics, and optics. They feature a high efficiency filtration system that provides a particle-free workstation around the manipulation, bringing HEPA filtered clean air down to the work surface. High efficiency UV lights are used to decontaminate the worktop from biological cross-contamination between operations.
The hoods come with Smart Technology™ , an exclusive set of tools that consists of Smart-Light Communication, chemical sensors (optional if adding carbon filters), real-time status, and the eGuard app. Smart Technology gives you an easily identifiable method of communication regarding the sash being opened while the UV light is on, air face velocity is compromised: check sash, pre-filter, or HEPA/ULPA, and fan status via a soft, LED band of light called Smart-Light. Smart-Light is a light signature across the front panel which casts a stable LED glow ensuring proper operation. If normal operation is disrupted, the reassuring LED light pulses and sounds, drawing the attention of the operator.
Units arrive completely broken down and will require assembly. Order replacement filter 33520-26, separately.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Hood Width (in) Power (VAC)
Availability Pricing
Erlab CaptairFlow 321 Clean Air  Enclosure with HEPA Filter and SS Work Surface, 32"; 115V 3352132 Mfr # FLOW321
COMPARE
53 32 115
Call For Price
Erlab CaptairFlow 321  Clean Air Enclosure with HEPA Filter and SS Work Surface, 32"; 230 V/50 Hz 3352134 Mfr # FLOW321 50HZ
COMPARE
53 32 230
Call For Price
Erlab CaptairFlow 391 Clean Air Enclosure with HEPA Filter and SS Work Surface, 39"; 115V 3352136 Mfr # FLOW391
COMPARE
53 39 115
Call For Price
Erlab CaptairFlow 391 Clean Air Enclosure with HEPA Filter and SS Work Surface, 39"; 230 V/50 Hz 3352138 Mfr # FLOW391 50HZ
COMPARE
53 39 230
Call For Price
Erlab CaptairFlow 714 Clean Air Enclosure with HEPA Filter and SS Work Surface, 71"; 115V 3352144 Mfr # FLOW714
COMPARE
60 71 115
Call For Price
Erlab CaptairFlow 714  Clean Air Enclosure with HEPA Filter and SS Work Surface, 71"; 230 V/50 Hz 3352146 Mfr # FLOW714 50HZ
COMPARE
60 71 230