Excel Scientific SealPlate Films for ELISA, EIA, and Microplates
Prevents evaporation and protects sample during processing, storage, and shipment
  • Superior quality film and uniform, consistently applied adhesive
  • Two removable end tabs for easy and accurate positioning on microplate
  • Functional temperature range from –40°C to 120°C
15,200.00 - ₹20,800.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (mm) Sterile Width (mm)
Availability Pricing
Excel Scientific TiterSeal Films, Non-Sterile; 100/PK 1302400 Mfr # TTS100
COMPARE
146.1 No 79.4
₹15,200.00INR / Pkg of 100
Excel Scientific SealPlate Films, Sterile; 100/Pk 1302402 Mfr # STR-SEAL-PLT
COMPARE
146.1 Yes 79.4
₹20,800.00INR / Pkg of 100