Excel Scientific ThinSeal Films for ELISA, Incubation, and Storage
Prevents evaporation and protects samples during processing, storage, and shipment
  • Superior quality film and uniform, consistently applied adhesive
  • Two removable end tabs for easy and accurate positioning on microplate
  • Functional temperature range from -40 to 120°C
ThinSeal™ films are just 25-µm thick, clearer and marginally piercable with a single-channel pipette or probe for sample recovery. Measure 79.4 x 141 mm.
18,900.00 - ₹25,300.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Sterile Width (mm)
Availability Pricing
Excel Scientific SealPlate ThinSeal Microplate Film, 25-μm Thick, Non-Sterile; 100/pk, Sterile 1302460 Mfr # 100-THIN-PLT
COMPARE
Polyester No 79.4
₹18,900.00INR / Pkg of 100
Excel Scientific SealPlate ThinSeal Microplate Film, 25-μm Thick, Sterile; 100/pk, Sterile 1302462 Mfr # STR-THIN-PLT
COMPARE
Polyester Yes 79.4
₹25,300.00INR / Pkg of 100