Filter Packages for Air Cleaning Systems
High-Efficiency Filters capture smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding or soldering. Prefilter is 30 to 35% efficient; high-efficiency cartridge is 96.5% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. Prefilter is 30 to 35% efficient; HEPA filter is 99.97% efficient.
Final Adsorption Carbon Module captures volatile solvents, touch-up paint fumes, and other noxious odors. Available in combination with primary high-efficiency or HEPA filter (33703-02 and -04), or use asa final stage for portable air cleaning system.
23,000.00 - ₹124,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Air Impurities Removal Systems Primary High-Efficiency Filter w/ Carbon Module for Air Cleaning System 3370302 Mfr # F-981-2A
COMPARE
Primary High-Efficiency Filter w/ Carbon Module for Air Cleaning System
₹109,000.00INR / Each
Air Impurities Removal Systems Primary HEPA Filter Package with Prefilter for Air Cleaning Systems 3370303 Mfr # F-981-3
COMPARE
Primary HEPA Filter Package with Prefilter for Air Cleaning Systems
₹96,300.00INR / Each
Air Impurities Removal Systems Primary HEPA Filter Package with Carbon Module for Air Cleaning Systems 3370304 Mfr # F-981-4A
COMPARE
Primary HEPA Filter Package with Carbon Module for Air Cleaning Systems
₹124,000.00INR / Each
Air Impurities Removal Systems Primary Final Adsorption Carbon Module for Air Cleaning Systems 3370305 Mfr # F-981-5A
COMPARE
Primary Final Adsorption Carbon Module for Air Cleaning Systems
₹112,000.00INR / Each
Air Impurities Removal Systems High-Efficiency Filter with Prefilter for Air Cleaning Systems 3370306 Mfr # RF-981-1
COMPARE
High-Efficiency Filter with Prefilter for Air Cleaning Systems
₹61,400.00INR / Each
Air Impurities Removal Systems Activated Carbon Refill for 33703-04, -05 3370307 Mfr # RF-981-2A
COMPARE
Activated Carbon Refill for 33703-04, -05
₹27,700.00INR / Each
Air Impurities Removal Systems Activated Carbon Refill for Air Cleaning Systems, fits 33703-02 3370308 Mfr # RF-981-2C
COMPARE
Activated Carbon Refill for Air Cleaning Systems, fits 33703-02
₹34,300.00INR / Each
Air Impurities Removal Systems HEPA Filter for Air Cleaning Systems 3370309 Mfr # RF-981-5
COMPARE
HEPA Filter for Air Cleaning Systems
₹83,700.00INR / Each
Air Impurities Removal Systems Prefilters for HEPA Filter for Air Cleaning Systems, Pack of Six 3370312 Mfr # RF_981-6
COMPARE
Prefilters for HEPA Filter for Air Cleaning Systems, Pack of Six
₹23,000.00INR / Pkg of 6
Air Impurities Removal Systems RF-400-1 Secondary Merv 8 Pleated Filter for Extract-All Systems 3370490 Mfr # RF-400-1
COMPARE
Secondary Merv 8 Pleated Filter for Extract-All Systems
₹27,400.00INR / Each
Air Impurities Removal Systems RF-400-2 Primary Merv 14 Pleated Filter for Extract-All Systems 3370491 Mfr # RF-400-2
COMPARE
Primary Merv 14 Pleated Filter for Extract-All Systems
Call For Price
Air Impurities Removal Systems RF-400-3 Activated Carbon After-Filter for Extract-All Systems 3370492 Mfr # RF-400-3
COMPARE
Activated Carbon After-Filter for Extract-All Systems
Call For Price
Showing 1- 12 of 12