Filter Packages for Compact Ductless Air Filter System
Easily install or change filters in just seconds. Filter packages designed to suit your specific working conditions
High-Efficiency Filters captures smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding. If you use solvents or paint in the same process, choose a combination high-efficiency filter with refillable 2" thick carbon module. Filter is 95% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. If you use solvents or paint in the same process, choose a combination HEPA filter with refillable 2" thick carbon module. Filter is 99.97% efficient.
Activated Carbon Filter captures volatile solvents, touch-up paint fumes, and other noxious odors. The primary
27,100.00 - ₹103,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Air Impurities Removal Systems Primary High-Efficiency Filter w/ Carbon Module for Ductless System 3370342 Mfr # F-987-2A
₹103,000.00INR / Each
COMPARE
Primary High-Efficiency Filter w/ Carbon Module for Ductless System
Loading
₹103,000.00INR / Each
Air Impurities Removal Systems Carbon Module with Final Filter for use with Ductless Air Filter System 3370345 Mfr # F-987-5A
₹99,000.00INR / Each
COMPARE
Carbon Module with Final Filter for use with Ductless Air Filter System
Loading
₹99,000.00INR / Each
Air Impurities Removal Systems Prefilters for Air Cleaning Systems, 4/pk 3370346 Mfr # RF-987-1
₹33,800.00INR / Pkg of 4
COMPARE
Prefilters for Air Cleaning Systems, 4/pk
Loading
₹33,800.00INR / Pkg of 4
Air Impurities Removal Systems Activated Carbon Refil for 33703-42, -44 3370348 Mfr # RF-987-4C
₹27,100.00INR / Each
COMPARE
Activated Carbon Refil for 33703-42, -44
Loading
₹27,100.00INR / Each
Air Impurities Removal Systems Final Filters for Ductless Air Filter Systems, 4/pk 3370349 Mfr # RF-987-6
₹42,200.00INR / Pkg of 4
COMPARE
Final Filters for Ductless Air Filter Systems, 4/pk
Loading
₹42,200.00INR / Pkg of 4