Foam Cleanroom Swabs
Ideal for cleaning small surface areas
  • Safely removes solvents and fluxes
  • Features low levels of nonvolatile residue
Foam swabs have sealed handles and are constructed from 100 pores per inch reticulated urethane foam — ideal for solvent dispersion and low-viscosity adhesives. Sealed foam swabs are perfect for removal of solvents and fluxes. Cleanfoam® swabs feature low levels of nonvolatile residue, particle generation, and are constructed of open-cell polyurethane foam for excellent solvent capacity.
8,100.00 - ₹32,100.00INR / Bag of 500
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compliance Swab Total Length (InchShort) Tip Length (InchShort)
Availability Pricing
Texwipe TX700B Cleanroom swab; foam bud tip and hardwood handle; 1.10"L tip, 500/pk 3367603 Mfr # TX700B
COMPARE
None 6 1/16 1 7/64
₹32,100.00INR / Bag of 500
Texwipe TX752B Cleanroom swab; Cleanfoam tip and polypropylene handle; 0.13"L tip 3367750 Mfr # TX752B
COMPARE
Class 100 2 3/4 1/8
₹24,500.00INR / Bag of 500
Texwipe TX741B Cleanroom swab; flexible Cleanfoam tip and polypropylene handle; 0.413"L tip 3367752 Mfr # TX741B
COMPARE
Class 100 2 3/4 13/32
₹17,300.00INR / Bag of 500
Texwipe TX740B Cleanroom swab; Cleanfoam tip and polypropylene handle; 0.67"L tip 3367755 Mfr # TX740B
COMPARE
Class 100 6 43/64
₹24,700.00INR / Bag of 500
Texwipe TX708A Cleanroom swab; circular-foam tip and polypropylene handle; 0.81"L tip 3367756 Mfr # TX708A
COMPARE
Class 100 5 13/64 13/16
₹8,100.00INR / Bag of 100
Texwipe TX750E Cleanroom Swab, Cleanfoam Tip and ESD Handle, 0.14"L Tip; 500/BG 3367772 Mfr # TX750E
COMPARE
Class 100 3 1/2 9/64
₹20,000.00INR / Bag of 500
Texwipe TX757E Cleanroom swab; Cleanfoam tip and ESD handle; 0.15"L tip 3367774 Mfr # TX757E
COMPARE
Class 100 2 3/4 5/32
₹19,100.00INR / Bag of 500
Showing 1- 7 of 7