Foxx Life Sciences Single Use Sterile Cap Assembly
Safely use your own bottles to transfer and store fluids in your work area
  • Create a closed system to collect fluids from your application
  • Create a system with ventilation or to prevent spills
  • Control flow into a container with the included pinch clamp
  • These assemblies are sterile and ready to use without the need to clean and validate
Single use cap assembly for connecting your bottles to your application and collect, transfer and store fluids. The production of our cap assembly is gamma irradiated and has no machining residue or bonding agents. Cap assemblies come with tubing, pinch clamp, and vent filter to fit an assortment of bottle sizes. Use these sterile cap assemblies right out of the bag with your application.
306,000.00 - ₹331,000.00INR / Case of 20
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fits Bottle / Beaker Size (mL) Neck Inside Diameter (ID) (mm) Overall Height (in)
Availability Pricing
Foxx Life Sciences Single Use Cap Assembly, 38-430, PP, Vented with Tube for 250 mL; 20/CS 0606134 Mfr # 430-0015-FLS
COMPARE
250 38 9 2/5
₹306,000.00INR / Case of 20
Foxx Life Sciences Single Use Cap Assembly, 38-430, PP, Vented with Tube for 1000 mL; 20/CS 0606136 Mfr # 430-0017-FLS
COMPARE
1000 38 12 19/64
₹331,000.00INR / Case of 20