General Tools Multi-Function Thermohygrometers
Compact and rugged design featuring a large display
  • Ideal for monitoring enviromental conditions
Measure temperature, humidity and dew point in a variety of applications including industrial, research, and building inspection These thermohygrometers simultaneously display any two parameters; temperature, humidity or dew point. View measurements in low light areas with backlit display. Meters can also be calibrated in the field using optional RH calibration standard salts (37803-05).
12,000.00 - ₹16,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
General Tools EP8703 Multi-Function Humidity Meter 3780476 Mfr # EP8703
COMPARE
1 9V (included) 99.9 122
₹12,000.00INR / Each
General Tools EP8709 Multi-Function Humidity Meter with Dew Point 3780477 Mfr # EP8709
COMPARE
1 9V (included) 99.9 122
₹16,900.00INR / Each
General Tools EP8710 Multi-Function Humidity Meter; Dewpoint, Remote Temp 3780478 Mfr # EP8710
COMPARE
1 9V (included) 99.9 122