GF+ Signet 9900 Single-Channel Universal Transmitter
Add-on modules help you create your own customized transmitter
  • Large, backlit LCD with ''Dial-type" digital bar graph
  • Customizable label and bar graph settings
  • User-friendly interface consistent with legacy instruments
  • Optional plug-in modules easily expand capibilities
These versatile, single-channel transmitters are designed to help you tailor a system to your exact needs. Transmitters feature a large LCD with digital bar graph as well as displays of the main and secondary measurements. Preset values offer quick and easy programming. Or customize your unit for labeling, min/max dial settings, and unit/decimal measurement options. Both panel- and field-mount models run on 12 to 32 VCD power and can be looped with compatible sensors.
For added flexibility, add-on modules let you create the ideal control center. The relay module 56560-15 (for panel mount transmitter only) features two programmable dry-contact relays for hysteresis and time delay. PC COMM module 56560-19 lets you easily configure your transmitter from your computer—save settings and use for future installations. Choose the H COMM module 56560-17 to communicate with any HART®-enabled device and remotely adjust 4 to 20 mA settings.
Expand your measurement capabilities using the optional parameter modules for applications including pH, ORP, temperature, flow, pressure, and level control. Contact our Application Specialists for details.
132,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GF Signet PANEL MOUNT Single-Channel Transmitter, Panel Mount 5656012 Mfr # 3-9900-1P
₹132,000.00INR / Each
COMPARE
Single-Channel Transmitter, Panel Mount
Loading
₹132,000.00INR / Each
GF Signet Single-Channel Transmitter, Field Mount 5656014 Mfr # 3-9900-1
₹132,000.00INR / Each
COMPARE
Single-Channel Transmitter, Field Mount
Loading
₹132,000.00INR / Each