Glove Box Replacement Gloves
Excellent glove box glove replacement
Order your replacement Hypalon, butyl, or neoprene gloves.
Butyl gloves are highly impermeable to water vapor, gases, and toxic chemicals.
Hypalon gloves are resistant to abrasions, ozone, and oxidizing chemicals.
Neoprene gloves are lightweight and embossed with firmhold finish.
All gloves are one-piece seamless, 0.015 gauge, come with O-rings, and fit 8 to 8 3/4" openings.
107,000.00 - ₹153,000.00INR / Pair of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Size
Availability Pricing
Labconco 5005500 Glove Box Gloves, Neoprene, Size 8.5, 30"L; 1 pair 0911212 Mfr # 5005500
COMPARE
30 Neoprene 8.5
₹107,000.00INR / Pair of 1
Labconco 5005600 Glove Box Gloves, Neoprene, Size 9.75, 30"L; 1 pair 0911213 Mfr # 5005600
COMPARE
30 Neoprene 9.75
₹107,000.00INR / Pair of 1
Labconco 5005501 Glove Box Gloves, Butyl, size 8.5, 30"L; 1 Pair 3476060 Mfr # 5005501
COMPARE
30 Butyl 8.5
₹153,000.00INR / Pair of 1
Labconco 5005601 Glove Box Gloves, Butyl, Size 9.75, 30"L; 1 Pair 3476061 Mfr # 5005601
COMPARE
30 Butyl 9.75
₹152,000.00INR / Pair of 1
Labconco 5005502 Glove Box Gloves, Hypalon, size 8.5, 30"L; 1 Pair 3476062 Mfr # 5005502
COMPARE
30 Hypalon® 8.5
₹125,000.00INR / Pair of 1
Labconco 5005602 Glove Box Gloves, Hypalon, Size 9.75, 30"L; 1 Pair 3476063 Mfr # 5005602
COMPARE
30 Hypalon® 9.75
₹125,000.00INR / Pair of 1