Grid-Top Bunsen Burners
Gas Burner distributes flame through 100 separate openings in stainless steel grid top for uniform heating. The grid diameter is 11⁄2" (3.8 cm). This burner gives twice the heat of a straight tube burner. This burner features a venturi tube for thorough mixing of gas and air, and adjustable gas orifice and air shutter for complete combustion. Adjustable flame size. Rustproof chrome-plated brass body with zinc-aluminum alloy base.
17,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
BTU Output (per hour) Gas Type Inlet Tube OD (in)
Availability Pricing
Humboldt H-5550 Accuflame Bunsen Burner, High-Temperature Burner, Natural Gas 3613020 Mfr # H-5550
COMPARE
1200 Natural Gas 0.4375
₹17,000.00INR / Each
Humboldt H-5600 Accuflame Bunsen Burner, High-Temperature Burner, Lp Gas 3613022 Mfr # H-5600
COMPARE
3200 Liquid Propane (LP) 0.4375
₹17,000.00INR / Each