Grieve DR Series Industrial Mechanical Convection Drawer Ovens
Ideal for drying small items requiring circulation of heated air
  • Calibrated hydraulic temperature controller
  • Interior surface is highly resistant to corrosion and oxidation
  • Incoloy® sheathed tubular heating elements
  • Eight separate stainless-steel drawers
These industrial mechanical convection drawer ovens are designed for individual processing or drying of plastic granules, powdered metal parts, and other small items requiring circulation of heated air. Eight separate stainless-steel drawers allow individual inspection of oven contents -- reduces heat loss and lowers operating costs associated with conventional oven doors. The bottom and sides of drawers have 3/4" dia perforations on 1 1/4" staggered centers.
The ovens feature Incoloy® sheathed tubular heating elements, a brushed stainless-steel exterior, and a stainless-steel interior which is highly resistant to corrosion and oxidation. Industrial rock wool insulation effectively retains heat for improved temperature uniformity. On-off panel switch controls power to the unit and a red pilot light in front of unit indicates on/off status of heating element. Other features include recirculating blower, calibrated hydraulic temperature controller, and silicone rubber door gasket.
The 115 VAC models include a 6-ft power cord with three-prong plug (no plug on 230 VAC models). Order the optional stand separately.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Max Temperature (° F) Power (VAC)
Availability Pricing
Grieve DR-350 Industrial Mechanical Convection Drawer Oven; 4.8 cu ft, 230V 5230215 Mfr # DR350-230V/60HZ
COMPARE
4.8 348.8 230