H-B Instrument Easy-Read Nonhazardous General-Purpose Glass Thermometers, Fahrenheit-scale
Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green"
  • Statement of accuracy included
These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications where the use of mercury or other hazardous liquids is not desired. Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. These thermometers come complete with ring top anti-roll fitting, individual serial number, statement of accuracy, recyclable packaging, and are RoHS compliant.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Overall Length (mm)
Availability Pricing
H-B Instrument B60301-0600 Easy-Read General Purpose Liquid-In-Glass Thermometer; 0 to 300F, 76mm Immersion, Environmentally Friendly 0800841 Mfr # 603010600
COMPARE
300 0 300
Call For Price
H-B Instrument B60301-0400 Easy-Read General Purpose Liquid-In-Glass Thermometer; -30 to 120F, 76mm Immersion, Environmentally Friendly 9031004 Mfr # 603010400
COMPARE
120 -30 300
Call For Price
H-B Instrument B60304-0300 Easy-Read General Purpose Liquid-In-Glass Thermometer; -30 to 120F, Total Immersion, Environmentally Friendly 9031005 Mfr # 603040300
COMPARE
120 -30 300