H-B Instrument Enviro-Safe Double-Safe Nonhazardous PFA-Coated Safety Glass Thermometers, Celsius-scale
PFA coating provides additional safety- no glass or contamination if the thermometer breaks
  • The filling and glass are contained within the PFA jacket, preventing contamination
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
Double-safe thermometers are PFA safety coated In the event of breakage the coating will help contain the broken liquid and glass. Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, ring top anti-roll fitting, individual serial numbered and have a statement of accuracy.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
H-B Instrument B60400-1200 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; -10/110C, Total Immersion, Environmentally Friendly 0807705 Mfr # 604001200
COMPARE
110 -10 200
Call For Price
H-B Instrument B60400-0100 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; -10/150C, 50mm Immersion, Environmentally Friendly 0807706 Mfr # 604000100
COMPARE
150 -10 200
Call For Price
H-B Instrument B60400-0600 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; -20/110C, 76mm Immersion, Environmentally Friendly 0807713 Mfr # 604000600
COMPARE
110 -20 300
Call For Price
H-B Instrument B60400-0700 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; -20/150C, 76mm Immersion, Environmentally Friendly 0807715 Mfr # 604000700
COMPARE
150 -20 300
Call For Price
H-B Instrument B60400-1800 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; -20/150C, Total Immersion, Environmentally Friendly 0807716 Mfr # 604001800
COMPARE
150 -20 300
Call For Price
H-B Instrument B60400-0500 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; -10/70C, 76mm Immersion, Environmentally Friendly 9030001 Mfr # 604000500
COMPARE
70 -10 300
Call For Price
H-B Instrument B60400-0800 Double-Safe PFA Coated, Liquid-In-Glass Thermometer; -10/260C, 76mm Immersion, Environmentally Friendly 9030003 Mfr # 604000800
COMPARE
260 -10 255
Call For Price
Showing 1- 7 of 7