H-B Instrument Standard Enviro-Safe Nonhazardous Pocket Glass Thermometers, Celsius-scale
Rugged case with an eyelet at the top for easy suspension
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
The case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation (except closed case styles). Available in three case styles; closed metal, open metal, or closed plastic. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant and have a statement of accuracy.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
H-B Instrument B60570-0200 Enviro-Safe Liquid-In-Glass Pocket Thermometer; -5 to 50C, Window Plastic Case, Environmentally Friendly 0800889 Mfr # 605700200
COMPARE
50 -5 160
Call For Price
H-B Instrument B60570-1900 Enviro-Safe Liquid-In-Glass Pocket Thermometer; -5 to 50C, Aluminum Duplex Case, Environmentally Friendly 0800893 Mfr # 605701900
COMPARE
50 -5 160