Idex® High-Pressure Stainless Steel Ferrules

Can withstand pressures up to 20,000 psi (1380 bar)!
  • Constructed of durable stainless steel
Use with IDEX high-pressure stainless steel nuts for your high-pressure applications. Stainless steel ferrules are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
910.00 - ₹8,900.00INR / Pkg of 10

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
Idex 316 Stainless Steel Ferrule, 1/16" OD Tubing, 10-32 Coned; 10/PK 0193990 Mfr # U-401X
COMPARE
Nuts 02022-36, -38, 39, -40, 02014-74, and 01939-80
₹8,900.00INR / Pkg of 10
Idex 316 Stainless Steel Ferrule, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK 0201469 Mfr # U-321X
COMPARE
Nut 02014-68
₹8,500.00INR / Pkg of 10
Idex 316 Stainless Steel Ferrule, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202235 Mfr # U-321
COMPARE
Nut 02014-68
₹930.00INR / Each
Idex 316 Stainless Steel Ferrule, 1/16" OD Tubing, 10-32/M6 Coned; 1/EA 0202237 Mfr # U-401
COMPARE
Nuts 02022-36, -38, 39, -40, 02014-74, and 01939-80
₹910.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat