Idex® Super Flangeless™ Ferrules, M6 Flat Bottom
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
1,700.00 - ₹17,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (mm) Fitting Material Description
Availability Pricing
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom; 1/EA 0202200 Mfr # P-357-2
COMPARE
2 PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom; 1/EA
₹2,100.00INR / Each
Idex Super Flangeless™ Ferrule Only, Natural PEEK, 2 mm OD Tubing, M6 Flat Bottom; 1/EA 0202201 Mfr # P-357-2-01
COMPARE
2 PEEK Super Flangeless™ Ferrule Only, Natural PEEK, 2 mm OD Tubing, M6 Flat Bottom; 1/EA
₹1,700.00INR / Each
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom; 10/PK 0202202 Mfr # P-357-2X
COMPARE
2 PEEK/SS Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom; 10/PK
₹17,800.00INR / Pkg of 10