Idex® Upchurch Stand-Alone Degassing Systems
Degas multiple solvent lines at the same time
  • Remove dissolved gasses continuously from mobile phases
  • Inert flow path
  • Low volume and easy to prime
These in-line full stand-alone degassing systems efficiently remove unwanted dissolved gases from mobile phases. Their unique design provides the highest level of reliable performance and makes helium sparging unnecessary. Optimal continuous operation is maintained by vacuum system monitoring.
385,000.00 - ₹784,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate Per Syringe (microL/min) Number of Channels
Availability Pricing
Idex Vacuum Degasser, 2 Channel, 480 µL Per Channel, 2 mL/min; 100 to 240 VAC  0201293 Mfr # 0001-6352
COMPARE
2 480 2
₹661,000.00INR / Each
Idex Vacuum Degasser, 4 Channel, 480 µL Per Channel, 2 mL/min; 100 to 240 VAC  0201294 Mfr # 0001-6354
COMPARE
2 480 4
₹784,000.00INR / Each
Idex Mini Lite Degasser, 1 channel, 285 µL/Channel, 2 mL/min; 100 to 240 VAC 4271240 Mfr # 0001-6682
COMPARE
2 285 1
₹385,000.00INR / Each