IKA® HRC 2 Series Refrigerated/Heating Circulating Baths
Powerful yet takes up only a small footprint—measures 8-3/4”W x 20-3/4”D (22 x 52.5 cm)
  • Powerful pressure/suction pump perfect for external heating
  • Large display for easy setting and viewing of parameters
  • RS-232 and USB interfaces—connect to a PC to control the circulators
  • Feed hopper and bleed valve offer safe and clean handling of the thermofluid
  • Easy access air filter makes cleaning convenient
Use HRC 2 refrigerated/heating circulator for tempering external devices such as bioreactors, viscometers, and polarimeters. Variable-speed pressure/suction pump offers powerful circulation yet operates at a quiet 52 dBA noise level.
PID temperature control provides excellent accuracy and stability to handle most demanding applications. The insulated 4-liter bath allows fast heating and reduces heat input when cooling. Connect the circulator to a PC via the USB or RS-232 interface; control and monitor with the labworldsoft® laboratory software. Fluid level indicator turns LED segment red if the fluid level is too high or low.
2,089,000.00 - ₹2,938,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
IKA HRC 2 basic Refrigerated/Heating Circulator, –20 to 100°C; 115V 2871177 Mfr # 25003743
COMPARE
100 –20 HRC 2 basic Refrigerated/Heating Circulator, –20 to 100°C; 115V
₹2,089,000.00INR / Each
IKA HRC 2 control Refrigerated/Heating Circulator with Wireless Controller, –30 to 100°C; 115V 2871178 Mfr # 25004639
COMPARE
100 –30 HRC 2 control Refrigerated/Heating Circulator with Wireless Controller, –30 to 100°C; 115V
₹2,938,000.00INR / Each