InnoCal® Mixer and Stirrer Calibrations
Quick and accurate mixer and stirrer calibrations accredited to ISO/IEC 17025 standards
  • NIST-traceable calibration allows you to confirm that your equipment meets the manufacturer’s published specification EPA, USDA, ISO and GLP/GMPs standards
  • Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
InnoCal provides calibration services for a wide variety of laboratory and industrial mixing and stirring equipment.
Stirrers and stirring hotplates can also be calibrated for rotational accuracy (rpm). All rotational motion (rpm) measurements are traceable to NIST and include data. Equipment is tested at 10 rpm values throughout the range. Accuracy is confirmed using NIST-traceable optical devices.
Service for stirring hotplates and hotplates adds NIST-traceable, temperature control system calibration.
Major brands of stirrers and stirring hotplates that InnoCal can service are Cole-Parmer, Corning, EchoTherm, Fisher Scientific Isotemp, IKA, Thermo Scientific (Cimarec, Super Nuova), Troemner.
43,100.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Test Points Calibration Type Description
Availability Pricing
COMPARE
Across the Range of the Instrument A2LA ISO17025 Accredited Temperature A2LA ISO17025 Accredited Calibration; Hot Plate Only
₹43,100.00INR / Each
COMPARE
Across the Range of the Instrument A2LA ISO17025 Accredited Temperature A2LA ISO17025 Accredited Calibration; Stirrer and Heat (Stirring Hot Plate)
₹43,100.00INR / Each
COMPARE
Across the Range of the Instrument A2LA ISO17025 Accredited Mechanical A2LA ISO17025 Accredited Calibration; Mixer
₹43,100.00INR / Each
COMPARE
Across the Range of the Instrument A2LA ISO17025 Accredited Mechanical A2LA ISO17025 Accredited Calibration; Stirrer Only
₹43,100.00INR / Each