Interscience BagMixer Paddle Blender Bag Stands and Racks

Specially designed bag holders and racks allow you to easily place samples in open bags.
35,100.00 - ₹101,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Interscience Laboratories 211-010 Bag Opener Stand For Holding Bags Size 80 And 100 mL, 1 Each 3685039 Mfr # 211-010
COMPARE
Bag Opener Stand For Holding Bags Size 80 And 100 mL, 1 Each
₹35,100.00INR / Each
Interscience Laboratories 211-040 Bag Opener Stand For Holding Bags Size 400 mL, 1 Each 3685040 Mfr # 211-040
COMPARE
Bag Opener Stand For Holding Bags Size 400 mL, 1 Each
₹35,100.00INR / Each
Interscience Laboratories 211-350 Bag Opener Stand For Holding Bags Size 3500 mL, 1 Each 3685041 Mfr # 211-350
COMPARE
Bag Opener Stand For Holding Bags Size 3500 mL, 1 Each
₹101,000.00INR / Each
Interscience Laboratories 221-010 Storage Racks For 11 Bags 80 And 100 mL, 1 Each 3685042 Mfr # 221-010
COMPARE
Storage Racks For 11 Bags 80 And 100 mL, 1 Each
₹53,900.00INR / Each
Interscience Laboratories 221-040 Storage Racks For 12 Bags 400 mL, 1 Each 3685043 Mfr # 221-040
COMPARE
Storage Racks For 12 Bags 400 mL, 1 Each
₹61,800.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat