Cole-Parmer® SP-200 Series Visible Spectrophotometers
Suited to a wide range of applications in education, quality control, environmental and clinical analysis
  • Wavelength range of 320 to 1000 nm
  • Absorbance, %T, and concentration modes
  • Analog and RS-232 outputs let you interface with printers and PCs
  • Multi-parameter display with wavelength and photometric readouts
  • Wide range of sampling accessories
The Cole-Parmer SP-200 visible spectrophotometers are high-quality, low-cost units perfect for schools/colleges and QC and testing labs. Models 83054-00 and 83054-07 with domed lid accept taller tubes with height of up to 105 mm.
481,000.00 - ₹984,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Wavelength (nm) Power (VAC) Description
Availability Pricing
COMPARE
1000 110 Visible Spectrophotometer; 110 VAC, 60 Hz
₹982,000.00INR / Each
COMPARE
1000 110 Visible Spectrophotometer with Domed Lid; 110 VAC, 60 Hz
₹481,000.00INR / Each
COMPARE
1000 230 Visible Spectrophotometer; 230 VAC, 50 Hz
₹984,000.00INR / Each
COMPARE
1000 230 Visible Spectrophotometer with Domed Lid; 230 VAC, 50 Hz
₹825,000.00INR / Each