Kimax Class B Burettes (Burets)
Graduations won’t wear off
  • Maintain fluid integrity within burette (buret) with dust cap
Manufactured from borosilicate glass, these funnel-fill Class B burettes (burets) are designed to meet ASTM E 287 requirements. Burettes feature durable black ceramic enamel graduations and a PTFE 2-mm bore stopcock plug.
43,700.00 - ₹55,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Tolerance
Availability Pricing
DWK Life Sciences (Kimble) Kimax Class B Burette, with PTFE stopcock, 25 mL, 1/each 3454203 Mfr # 17026F-25
COMPARE
25 0.1 0.06
₹43,700.00INR / Each
DWK Life Sciences (Kimble) Kimax Class B Burette, with PTFE stopcock, 50 mL, 1/each 3454205 Mfr # 17026F-50
COMPARE
50 0.1 0.1
₹47,700.00INR / Each
DWK Life Sciences (Kimble) 17026F-100 Class B Burette, with PTFE stopcock, 100 mL, 1/each 3454207 Mfr # 17026F-100
COMPARE
100 0.2 0.2
₹55,200.00INR / Each