KIMAX® Color-Coded Glass Griffin Beakers
Get quick and easy identification with brightly colored markings
Autoclavable
  • Permanent markings won’t wear off
These clear, color-colored Griffin beakers are made of Kimax KG-33 glass for strength and durability. Beakers have uniform sidewall and bottom thicknesses, and feature a color matte finish marking area for use with an ordinary pencil. Each has permanent, easy-to-read color graduations. Use beakers up to maximum temperature of 840°F (450°C).
Reference: ASTM E-960, Type I.
14,200.00 - ₹28,300.00INR / Case of 12
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
DWK Life Sciences (Kimble) 14000B-50 blue-coded Griffin beakers; 50 mL, 12/cs 3451500 Mfr # 14000B-50
COMPARE
50 10 Glass
₹20,800.00INR / Case of 12
DWK Life Sciences (Kimble) 14000B-100 blue-coded Griffin beakers; 100 mL, 12/cs 3451502 Mfr # 14000B-100
COMPARE
100 10 Glass
₹20,800.00INR / Case of 12
DWK Life Sciences (Kimble) 14000B-150 blue-coded Griffin beakers; 150 mL, 12/cs 3451504 Mfr # 14000B-150
COMPARE
150 20 Glass
₹20,100.00INR / Case of 12
DWK Life Sciences (Kimble) 14000B-250 blue-coded Griffin beakers; 250 mL, 12/cs 3451506 Mfr # 14000B-250
COMPARE
250 25 Glass
₹20,100.00INR / Case of 12
DWK Life Sciences (Kimble) 14000B-400 blue-coded Griffin beakers; 400 mL, 12/cs 3451508 Mfr # 14000B-400
COMPARE
400 25 Glass
₹22,300.00INR / Case of 12
DWK Life Sciences (Kimble) 14000B-600 blue-coded Griffin beakers; 600 mL, 6/cs 3451510 Mfr # 14000B-600
COMPARE
600 50 Glass
₹14,200.00INR / Case of 6
DWK Life Sciences (Kimble) 14000B-1000 blue-coded Griffin beakers; 1000 mL, 6/cs 3451512 Mfr # 14000B-1000
COMPARE
1000 50 Glass
₹28,300.00INR / Case of 6
DWK Life Sciences (Kimble) 14000Y-250 yellow-coded Griffin beakers; 250 mL, 12/cs 3451534 Mfr # 14000Y-250
COMPARE
250 25 Glass
₹15,400.00INR / Case of 12
DWK Life Sciences (Kimble) 14000Y-400 yellow-coded Griffin beakers; 400 mL, 12/cs 3451536 Mfr # 14000Y-400
COMPARE
400 25 Glass
Call For Price
Showing 1- 9 of 9