KIMAX® Erlenmeyer Flasks
Erlenmeyer, narrow mouth, with graduations
  • Narrow mouth with graduations
These versatile flasks are ideal for general laboratory measuring and mixing where precision accuracy is not required. The thick wall and tapered design provide strength and minimize chipping. Tops are tooled with rounded edges. Approximate graduations are in durable white enamel. Compliance: ASTM E1404, type I, class 1
65,500.00 - ₹79,400.00INR / Case of 48
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Stopper Number Accepted
Availability Pricing
DWK Life Sciences (Kimble) 26500-125 Erlenmeyer Glass Flask, 125 mL, stopper size 5, 48/cs 3452414 Mfr # 26500-125
COMPARE
125 Glass 5
₹79,400.00INR / Case of 48
DWK Life Sciences (Kimble) 26500-2000 Erlenmeyer Glass Flask, 2000 mL, stopper size 10, 8/cs 3452424 Mfr # 26500-2000
COMPARE
2000 Glass 10
₹65,500.00INR / Case of 8