Kimble Separatory Funnels
Meets ASTM specification E1096, Type 4 requirements
  • PTFE stopcocks
Funnel is manufactured with stem ID large enough that allows separation when stopcock is closed; allowing a more complete separation and eliminates the need for emptying the funnel.
132,000.00 - ₹249,000.00INR / Case of 4
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Stopcock Plug Size
Availability Pricing
DWK Life Sciences (Kimble) 29048F30 Separatory Funnel, PTFE Stopcock, 30 mL 4/cs 3450540 Mfr # 29048F-30
COMPARE
30 Glass 2
₹132,000.00INR / Case of 4
DWK Life Sciences (Kimble) 29048F60 Separatory Funnel, PTFE Stopcock, 60 mL 4/cs 3450542 Mfr # 29048F60
COMPARE
60 Glass 2
₹169,000.00INR / Case of 4
DWK Life Sciences (Kimble) 29048F125 Separatory Funnel, PTFE Stopcock, 125 mL 4/cs 3450544 Mfr # 29048F125
COMPARE
125 Glass 2
₹163,000.00INR / Case of 4
DWK Life Sciences (Kimble) 29048F250 Separatory Funnel, PTFE Stopcock, 250 mL 4/cs 3450546 Mfr # 29048F250
COMPARE
250 Glass 4
₹193,000.00INR / Case of 4
DWK Life Sciences (Kimble) 29048F500 Separatory Funnel, PTFE Stopcock, 500 mL 4/cs 3450548 Mfr # 29048F500
COMPARE
500 Glass 4
₹222,000.00INR / Case of 4
DWK Life Sciences (Kimble) 29048F1000 Separatory Funnel, PTFE Stopcock, 1000 mL 2/cs 3450550 Mfr # 29048F1000
COMPARE
1000 Glass 4
₹153,000.00INR / Case of 2
DWK Life Sciences (Kimble) 29048F2000 Separatory Funnel, PTFE Stopcock, 2000 mL 2/cs 3450552 Mfr # 29048F2000
COMPARE
2000 Glass 6
₹249,000.00INR / Case of 2
Showing 1- 7 of 7