Kimcote, Class A, Graduated Cylinders with Bumpers
  • Calibrated “to deliver”
Ultra-clear safety-coated glassware helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware has a unique nonslip surface when wet or dry.Class A graduated cylinders are unique with reverse metric scale with white markings. Cylinders include a bumper and a ground base for stability. Autoclavable to120°C (250°F).
25,800.00 - ₹99,900.00INR / Case of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
DWK Life Sciences (Kimble) KC20028W-100 Class A, Graduated Cylinders with Bumpers, 100 mL 3452744 Mfr # KC20028W-100
COMPARE
100 1 Glass
₹25,800.00INR / Case of 1
DWK Life Sciences (Kimble) KC20028W-250 Class A, Graduated Cylinders with Bumpers, 250 mL 3452746 Mfr # KC20028W-250
COMPARE
250 2 Glass
₹34,000.00INR / Case of 1
DWK Life Sciences (Kimble) KC20028W-500 Class A, Graduated Cylinders with Bumpers, 500 mL 3452748 Mfr # KC20028W-500
COMPARE
500 5 Glass
₹50,800.00INR / Case of 1
DWK Life Sciences (Kimble) KC20028W-1000 Class A, Graduated Cylinders with Bumpers, 1000 mL 3452750 Mfr # KC20028W-1000
COMPARE
1000 10 Glass
₹57,500.00INR / Case of 1
DWK Life Sciences (Kimble) KC20028W-2000 Class A, Graduated Cylinders with Bumpers, 2000 mL 3452752 Mfr # KC20028W-2000
COMPARE
2000 20 Glass
₹99,900.00INR / Case of 1